Cæsiumsulfat ≥99%
Glass bottle for solids, 500 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Purity: 99% CAS No.: 10294-54-9 Molecular Formula: Cs2SO4 Formula Weight: 361.87 Odour: Odourless Melting Point: 1010° MDL No.: MFCD00010959
- CAS nummer:10294-54-9
- Formel:Cs₂O₄S
- MDL nummer:MFCD00010959
- Molecular Weight:361.87 g/mol
- Merck Index:14,2016
- Kogende Pt:1627 °C (1013 hPa)
- Smeltende Pt:1010 °C
- Tæthed:4,24
- Opbevaringstemperatur:Stue temperatur
- EINECS:233-662-6
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:500 g



