Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Cæsiumsulfat ≥99%

Cæsiumsulfat ≥99%

Glass bottle for solids, 100 g
Katalog # A16767.22
Leverandør:   Thermo Fisher Scientific
Cæsiumsulfat ≥99%
Katalog # A16767.22
CAS nummer:  10294-54-9
Cæsiumsulfat ≥99%
Cæsiumsulfat ≥99%

Glass bottle for solids, 100 g

CAS nummer:  10294-54-9
Price
1.300,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 10294-54-9 Molecular Formula: Cs2SO4 Formula Weight: 361.87 Odour: Odourless Melting Point: 1010° MDL No.: MFCD00010959

  • CAS nummer:10294-54-9
  • Formel:Cs₂O₄S
  • MDL nummer:MFCD00010959
  • Molecular Weight:361.87 g/mol
  • Merck Index:14,2016
  • Kogende Pt:1627 °C (1013 hPa)
  • Smeltende Pt:1010 °C
  • Tæthed:4,24
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:233-662-6
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.