Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumdecylsulfat ≥99%

Natriumdecylsulfat ≥99%

Glass bottle for solids, 1 g
Katalog # A16299.03
Ætsende
Irriterende
Leverandør:   Thermo Fisher Scientific
Natriumdecylsulfat ≥99%
Katalog # A16299.03
CAS nummer:  142-87-0
Natriumdecylsulfat ≥99%
Natriumdecylsulfat ≥99%

Glass bottle for solids, 1 g

CAS nummer:  142-87-0
Price
539,00 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents

Purity: 99% CAS No.: 142-87-0 Molecular Formula: CH3(CH2)9OSO3Na Formula Weight: 260.33 Beilstein Reference: 3576860 MDL No.: MFCD00041881

Synonymer:  Decyl sodium sulphate, Decyl sodium sulfate, Decyl sulphuric acid salt, Decyl sulfuric acid salt
  • CAS nummer:142-87-0
  • Formel:C₁₀H₂₁NaO₄S
  • MDL nummer:MFCD00041881
  • Molecular Weight:260.33 g/mol
  • Smeltende Pt:191,85 °C
  • Tæthed:1,207 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:205-568-5
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 g

Specifikationer