Orders LinkContactUsLinkComponent
Clamps for universal platforms
Clamps for universal platforms
Klemme til mikrotiterplade/dybbrøndsplade, 5 (280×330); 10 (457×457); 15 (457×609) for MaxQ 2000
18 (760×457) and 29 (910×609) clamps for MaxQ 3000
Katalog # 444-0002
Leverandør:   Thermo Fisher Scientific
Clamps for universal platforms
Katalog # 444-0002
Clamps for universal platforms
Clamps for universal platforms
Klemme til mikrotiterplade/dybbrøndsplade, 5 (280×330); 10 (457×457); 15 (457×609) for MaxQ 2000
18 (760×457) and 29 (910×609) clamps for MaxQ 3000
Price
312,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Documents
Specifikationer
  • Til:
    5 (280×330); 10 (457×457); 15 (457×609) for MaxQ 2000
    18 (760×457) and 29 (910×609) clamps for MaxQ 3000
  • Beskrivelse:
    Klemme til mikrotiterplade/dybbrøndsplade