Clamps for universal platforms
Klemme til 2800 ml Fernbach-kolbe, 1 (280×330); 4 (457×457); 4 (457×609) for MaxQ 2000
5 (760×457) and 8 (910×609) clamps for MaxQ 3000
5 (760×457) and 8 (910×609) clamps for MaxQ 3000
Katalog # 444-0316
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Documents
BARN30156BI_TDS_EN
Datablad
Specifikationer
- Til:1 (280×330); 4 (457×457); 4 (457×609) for MaxQ 2000
5 (760×457) and 8 (910×609) clamps for MaxQ 3000 - Beskrivelse:Klemme til 2800 ml Fernbach-kolbe



