Clamps for universal platforms
Klemme til lav 2500 ml celledyrkningskolbe, 1 (280×330); 1 (457×457); 2 (457×609) for MaxQ 2000
3 (760×457) and 6 (910×609) clamps for MaxQ 3000
3 (760×457) and 6 (910×609) clamps for MaxQ 3000
Katalog # 444-0315
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Documents
BARN30156BI_TDS_EN
Datablad
Specifikationer
- Til:1 (280×330); 1 (457×457); 2 (457×609) for MaxQ 2000
3 (760×457) and 6 (910×609) clamps for MaxQ 3000 - Beskrivelse:Klemme til lav 2500 ml celledyrkningskolbe
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



