Orders LinkContactUsLinkComponent
DNA ladders, SimplyLoad™
DNA ladders, SimplyLoad™
SimplyLoad™ Tandem DNA ladder: 60 ng/µl, 21 fragments from 100 bp to 12,000 bp, in 100 bp and 1,000 bp increments (bands at 1 kb and 5 kb stain brighter to aid identification), 1
Katalog # LONZ50333
Leverandør:   LONZA
DNA ladders, SimplyLoad™
Katalog # LONZ50333
Leverandør:  LONZA
CAS nummer:  
DNA ladders, SimplyLoad™
DNA ladders, SimplyLoad™
SimplyLoad™ Tandem DNA ladder: 60 ng/µl, 21 fragments from 100 bp to 12,000 bp, in 100 bp and 1,000 bp increments (bands at 1 kb and 5 kb stain brighter to aid identification), 1
CAS nummer:  
Price
2.880,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Lonza SimplyLoad™ DNA ladders are ready to load directly onto the gel to maximise time and convenience. Each band has an ideal 50% G-C content for rapid, precise sizing and sharp resolution. Free from contaminating plasmid DNA so they have minimal background.
  • Ready to load
  • Storage temperature: 2 to 8 °C
Specifikationer
Specifikationer
  • Pakn:
    1
  • Beskrivelse:
    SimplyLoad™ Tandem DNA ladder: 60 ng/µl, 21 fragments from 100 bp to 12,000 bp, in 100 bp and 1,000 bp increments (bands at 1 kb and 5 kb stain brighter to aid identification)
Specifikationer