Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Buffer solutions in sachets, Certipur®, Supelco®

Buffer solutions in sachets, Certipur®, Supelco®

Certipur® buffer solution, pH 11,00, boric acid, sodium hydroxide, potassium chloride, 11.00, Borat, Sachet, 30×30 ml, 30
Catalog # 1.99021.0001
Supplier:   Merck
Buffer solutions in sachets, Certipur®, Supelco®
Catalog # 1.99021.0001
Supplier:  Merck
CAS Number:  
Buffer solutions in sachets, Certipur®, Supelco®
Buffer solutions in sachets, Certipur®, Supelco®

Certipur® buffer solution, pH 11,00, boric acid, sodium hydroxide, potassium chloride, 11.00, Borat, Sachet, 30×30 ml, 30

CAS Number:  
Price
868,00 kr
Log in for availability
1 * 30 Items
Product Details & Documents

Til kalibrering af pH-metre.

  • Practical 30 ml sachets - one use per sachet - no risk of contamination
  • Solutions are always fresh and ready to use
  • Traceable to SRM from NIST and PTB
  • Reference temperature: +25 °C
  • Precision: Deviation is max. ±0,015 pH units

pH value is measured with a combined glass electrode after 5-point calibration according to DIN 19268 with reference buffer solutions according to DIN 19266, IUPAC, NIST, Ph. Eur. and USP.

Specifications

Specifications

  • Pakning:
    Sachet
  • pH buffer type:
    Borat
  • Pakn:
    30
  • Beskrivelse:
    Certipur® buffer solution, pH 11,00, boric acid, sodium hydroxide, potassium chloride
  • Pk Info:
    30×30 ml
  • pH-værdi:
    11.00
  • Cat. No.:
    1.99021.0001
  • M-Clarity quality level:
    MQ100

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.