Buffer solutions in sachets, Certipur®, Supelco®
Certipur® buffer solution, pH 2,00, citric acid, sodium hydroxide, hydrogen chloride, 2,00, Citrat, Sachet, 30×30 ml, 30
: Merck
Til kalibrering af pH-metre.
- Practical 30 ml sachets - one use per sachet - no risk of contamination
- Solutions are always fresh and ready to use
- Traceable to SRM from NIST and PTB
- Reference temperature: +25 °C
- Precision: Deviation is max. ±0,015 pH units
pH value is measured with a combined glass electrode after 5-point calibration according to DIN 19268 with reference buffer solutions according to DIN 19266, IUPAC, NIST, Ph. Eur. and USP.
Documents
- Pakning:Sachet
- pH buffer type:Citrat
- Pakn:30
- Beskrivelse:Certipur® buffer solution, pH 2,00, citric acid, sodium hydroxide, hydrogen chloride
- Pk Info:30×30 ml
- pH-værdi:2,00
- Cat. No.:1.99012.0001
- M-Clarity quality level:MQ100
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



