Assortment Boxes with Lid
with 18 compartments, 230×335×50 mm
Katalog # SARS95.958
Marketsource
Leverandør: SARSTEDT
Gå til:
Product Details & Documents
These assortment boxes are made of high quality impact resistant plastic with green base and transparent lid.
Specifikationer
- Beskrivelse:with 18 compartments
- B×D×H:230×335×50 mm