Tip system box and empty reload trays
Empty reload tray, for 96×200 µl pipette tips, gul
SARS95.1760.011
Marketsource
: SARSTEDT
The tip box with hinged lid is the central workstation for all 96-tip Sarstedt trays.
: Empty pipette tip trays need to be ordered separately.
- Farve:gul
- Beskrivelse:Empty reload tray, for 96×200 µl pipette tips
- Seq:05