Transport Cooling Containers
Styrofoam casing with rubber band
Katalog # SARS95.1011
Marketsource
Leverandør: SARSTEDT
Gå til:
Product Details & Documents
The cool transport containers is suitable for safe cool transportation of medical and biological specimens. It can accommodate two sample tubes of 10 ml volume or any tubes of 16,5 mm Ø and 107 mm length.
The mailing container is filled with a coolant and should be frozen in a horizontal position at −12 °C before mailing. The styrofoam casing protects against warmth and any damage during transport. The container is not suitable for samples that are not allowed to freeze.
Specifikationer
- Beskrivelse:Styrofoam casing with rubber band
- Seq:02