Slide mailers
Slide mailers with ventilation, hvid, PP, 2/4
Katalog # SARS86.972
Marketsource
Leverandør: SARSTEDT
Gå til:
Product Details & Documents
These mailing vessels are manufactured from polypropylene and is suitable for the safe transportation or storage of two glass microscope slides (or four if placed against each other).
- The lid features a ventilation hole
Bestillingsoplysninger: Containers available with or without ventilation hole.
Emballering: 50 pieces per sachet.
Specifikationer
- Antal objektglas:2/4
- Farve:hvid
- Beskrivelse:Slide mailers with ventilation
- Materiale:PP
- Seq:02