Orders LinkContactUsLinkComponent
Tissue culture flasks
TC-kolbe, standardoverflade, 250 ml, 75 cm², Skrå, 250, 5/pose, 100/kasse
  SARS83.1813.001
Marketsource
 :   SARSTEDT
Tissue culture flasks
  SARS83.1813.001
 :  SARSTEDT
Marketsource
Tissue culture flasks
TC-kolbe, standardoverflade, 250 ml, 75 cm², Skrå, 250, 5/pose, 100/kasse
1 * 100 Items
High-quality polystyrene.
  • Sterilised by gamma irradiation
  • Hydrophilic surface for optimal attachment and growth of adherent cells
  • Anti-tip skirt at canted neck
With red PE quick release (with or without ventilation) screw cap or black PP rim seal cap.
  • Dyrkningsareal:
    75 cm²
  • Pakket:
    5/pose, 100/kasse
  • Anbefalet arbejdsvolumen:
    250
  • Kapacitet:
    250 ml
  • Beskrivelse:
    TC-kolbe, standardoverflade
  • Hals:
    Skrå