96-Well micro test plates
Mikrotestplade med U-bund, uden låg, klar, 96, 25 stk./pose
SARS82.1582
Marketsource
: SARSTEDT
Clear PS
- Compatible with all common microplate dispensers, readers, and washers
- Excellent and consistent optical quality
- Optimal surface
- Uniform design
- Choice of flat, round or conical wells
- Ant. brønde:96
- Pakket:25 stk./pose
- Farve:klar
- Beskrivelse:Mikrotestplade med U-bund, uden låg