Stirring rods
Stirring rod, HDPE, 3 mm, 64 mm
Katalog # SARS81.971
Marketsource
Leverandør: SARSTEDT
Gå til:
Product Details & Documents
Stirring rods.
Specifikationer
- Ø:3 mm
- Beskrivelse:Stirring rod
- Længde:64 mm
- Materiale:HDPE
- Seq:02