Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Sample cups for Cobas system

Sample cups for Cobas system

For Cobas systems Bio, Mia and Fara, with looped cap, 0,6 ml, 34 mm, 8,2 mm, grøn
Katalog # SARS73.910.002
Marketsource
Leverandør:   SARSTEDT
Sample cups for Cobas system
Katalog # SARS73.910.002
Leverandør:  SARSTEDT
Marketsource
Sample cups for Cobas system

For Cobas systems Bio, Mia and Fara, with looped cap, 0,6 ml, 34 mm, 8,2 mm, grøn

Price
Spørg for pris
Log ind for tilgængelighed
1 * 5.000 Items
Product Details & Documents

Sample cups for Cobas system such as Bio, Mira, Fara and Integra.

  • Good chemical resistance
  • Good temperature resistance

Cups for Integra has cap with hole to insert its probe.

Specifikationer
  • Udv.Ø:
    8,2 mm
  • Kapacitet:
    0,6 ml
  • Farve:
    grøn
  • Beskrivelse:
    For Cobas systems Bio, Mia and Fara, with looped cap
  • Højde:
    34 mm
  • Seq:
    03
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.