Orders LinkContactUsLinkComponent
Microtubes
Microtubes
Tubes with screw cap non assembled, 2,0 ml, 5000/case, 2,0 ml
  SARS72.693.100
Marketsource
 :   SARSTEDT
Microtubes
  SARS72.693.100
 :  SARSTEDT
Marketsource
Microtubes
Tubes with screw cap non assembled, 2,0 ml, 5000/case, 2,0 ml
1 * 5.000 Items
PP, translucent
  • Excellent chemical resistance
  • Excellent temperature resistance
Without or with skirted base.
Documents
  • Pakket:
    5000/case
  • Anbefalet arbejdsvolumen:
    2,0 ml
  • Kapacitet:
    2,0 ml
  • Beskrivelse:
    Tubes with screw cap non assembled
Accessories
  •  
    Beskrivelse
  • SARS65.713.006
    Colour coded inserts, blue
    1 * 1.000 Items
  •  
    Beskrivelse
  • SARS65.712
    Screw cap with seperate retaining loop, neutral
    1 * 5.000 Items
  •  
    Beskrivelse
  • SARS65.713
    Colour coded inserts, neutral
    1 * 5.000 Items
  •  
    Beskrivelse
  • SARS65.713.002
    Colour coded inserts, red
    1 * 5.000 Items
  •  
    Beskrivelse
  • SARS65.713.007
    Colour coded inserts, violet
    1 * 1.000 Items
  •  
    Beskrivelse
  • SARS65.713.009
    Colour coded inserts, white
    1 * 1.000 Items
  •  
    Beskrivelse
  • SARS65.713.010
    Colour coded inserts, black
    1 * 5.000 Items
  •  
    Beskrivelse
  • SARS65.713.004
    Colour coded inserts, yellow
    1 * 5.000 Items
  •  
    Beskrivelse
  • SARS65.713.005
    Colour coded inserts, green
    1 * 5.000 Items