Microtubes with thread in special sterile packaging
Microcentrifuge tubes without skirted base, without knurls, sterile, 1,5 ml, 500/double bag, 2500/case, 1,5
Katalog # SARS72.687.772
Marketsource
Leverandør: SARSTEDT
Microtubes with thread in special sterile packaging
Katalog # SARS72.687.772
Leverandør: SARSTEDT
Marketsource
Gå til:
Product Details & Documents
PP, translucent
- With or without skirted base
Each bag is sub-packaged into a second additional bag. Removing the outer bag leaves a complete and sterile bag packaging for convenient processing in clean rooms or sterile environments. This packaging option is ideal for users working under clean room conditions to fill precious reagents into tubes.
Specifikationer
- Pakket:500/double bag, 2500/case
- Anbefalet arbejdsvolumen:1,5
- Kapacitet:1,5 ml
- Beskrivelse:Microcentrifuge tubes without skirted base, without knurls, sterile
Accessories
Produktinformation
- Katalog #TilFarveTilgængelighedPrice
- Colour coded screw caps, PP, sterileColour coded screw caps, PP, sterileSARS65.716.720Micro tubesgulLog ind for tilgængelighed1 * 5.000 Items
Produktinformation
- Katalog #TilFarveTilgængelighedPrice
- Colour coded screw caps, PP, sterileColour coded screw caps, PP, sterileSARS65.716.721Micro tubesrødLog ind for tilgængelighed1 * 5.000 Items
Produktinformation
- Katalog #TilFarveTilgængelighedPrice
- Colour coded screw caps, PP, sterileColour coded screw caps, PP, sterileSARS65.716.723Micro tubesblåLog ind for tilgængelighed1 * 5.000 Items - 1.836,66 kr
Produktinformation
- Katalog #TilFarveTilgængelighedPrice
- Colour coded screw caps, PP, sterileColour coded screw caps, PP, sterileSARS65.716.725Micro tubesneutraltLog ind for tilgængelighed1 * 5.000 Items - 1.922,91 kr
Produktinformation
- Katalog #TilFarveTilgængelighedPrice
- Colour coded screw caps, PP, sterileColour coded screw caps, PP, sterileSARS65.716.726Micro tubesgrønLog ind for tilgængelighed1 * 5.000 Items
Produktinformation
- Katalog #TilFarveTilgængelighedPrice
- Colour coded screw caps, PP, sterileColour coded screw caps, PP, sterileSARS65.716.727Micro tubesorangeLog ind for tilgængelighed1 * 5.000 Items
Produktinformation
- Katalog #TilFarveTilgængelighedPrice
- Colour coded screw caps, PP, sterileColour coded screw caps, PP, sterileSARS65.716.729Micro tubesbrunLog ind for tilgængelighed1 * 5.000 Items
Produktinformation
- Katalog #TilFarveTilgængelighedPrice
- Colour coded screw caps, PP, sterileColour coded screw caps, PP, sterileSARS65.716.728Micro tubessortLog ind for tilgængelighed1 * 5.000 Items - 1.922,91 kr
Produktinformation
- Katalog #TilFarveTilgængelighedPrice
- Colour coded screw caps, PP, sterileColour coded screw caps, PP, sterileSARS65.716.757Micro tubeshvidLog ind for tilgængelighed1 * 5.000 Items - 1.922,91 kr
Produktinformation
- Katalog #TilFarveTilgængelighedPrice
- Colour coded screw caps, PP, sterileColour coded screw caps, PP, sterileSARS65.716.755Micro tubesVioletLog ind for tilgængelighed1 * 5.000 Items