Orders LinkContactUsLinkComponent
Microtubes with thread in special sterile packaging
Microcentrifuge tubes with skirted base, sterile, 2,0 ml, 500/double bag, 2500/case
  SARS72.609.711
Marketsource
 :   SARSTEDT
Microtubes with thread in special sterile packaging
  SARS72.609.711
 :  SARSTEDT
Marketsource
Microtubes with thread in special sterile packaging
Microcentrifuge tubes with skirted base, sterile, 2,0 ml, 500/double bag, 2500/case
1 * 2.500 Items
PP, translucent
  • With or without skirted base
Each bag is sub-packaged into a second additional bag. Removing the outer bag leaves a complete and sterile bag packaging for convenient processing in clean rooms or sterile environments. This packaging option is ideal for users working under clean room conditions to fill precious reagents into tubes.
  • Pakket:
    500/double bag, 2500/case
  • Kapacitet:
    2,0 ml
  • Beskrivelse:
    Microcentrifuge tubes with skirted base, sterile
Accessories
  •  
    Beskrivelse
    Til
    Farve
  • SARS65.716.720
    Colour coded screw caps, PP, sterile
    Micro tubes
    gul
    1 * 5.000 Items
  •  
    Beskrivelse
    Til
    Farve
  • SARS65.716.721
    Colour coded screw caps, PP, sterile
    Micro tubes
    rød
    1 * 5.000 Items
  •  
    Beskrivelse
    Til
    Farve
  • SARS65.716.723
    Colour coded screw caps, PP, sterile
    Micro tubes
    blå
    1 * 5.000 Items - 1.836,66 kr
  •  
    Beskrivelse
    Til
    Farve
  • SARS65.716.725
    Colour coded screw caps, PP, sterile
    Micro tubes
    neutralt
    1 * 5.000 Items - 1.922,91 kr
  •  
    Beskrivelse
    Til
    Farve
  • SARS65.716.726
    Colour coded screw caps, PP, sterile
    Micro tubes
    grøn
    1 * 5.000 Items
  •  
    Beskrivelse
    Til
    Farve
  • SARS65.716.727
    Colour coded screw caps, PP, sterile
    Micro tubes
    orange
    1 * 5.000 Items
  •  
    Beskrivelse
    Til
    Farve
  • SARS65.716.729
    Colour coded screw caps, PP, sterile
    Micro tubes
    brun
    1 * 5.000 Items
  •  
    Beskrivelse
    Til
    Farve
  • SARS65.716.728
    Colour coded screw caps, PP, sterile
    Micro tubes
    sort
    1 * 5.000 Items - 1.922,91 kr
  •  
    Beskrivelse
    Til
    Farve
  • SARS65.716.757
    Colour coded screw caps, PP, sterile
    Micro tubes
    hvid
    1 * 5.000 Items - 1.922,91 kr
  •  
    Beskrivelse
    Til
    Farve
  • SARS65.716.755
    Colour coded screw caps, PP, sterile
    Micro tubes
    Violet
    1 * 5.000 Items