Pipette tips
Type L, 300 µl, +, Transparent, Eppendorf Reference, Research/Research Plus, mLine Mechanical, Proline Mechanical, eLine Electronic, 96/box, 480/inner case, 1920/case
Katalog # SARS70.765.200
Marketsource
Leverandør: SARSTEDT
Gå til:
Product Details & Documents
These high quality tips are suitable for the most common pipettes.
- Sterile tips are DNase/RNase- and ATP-free, and non pyrogenic
- Available loose in bags, packed in racks or in Tip StackPack reload systems
Tip StackPack: Tips are preloaded in reusable, stacked 96-tip trays. The trays are designed to safely cover each individual 96-tip layer, shielding the preloaded tips in the stack like a protective screen.
Specifikationer
- Pakket:96/box, 480/inner case, 1920/case
- Steril:+
- Volumen:300 µl
- Seq:24
- Spidstype:Type L
- Spits farge:Transparent
- Til brug med:Eppendorf Reference, Research/Research Plus, mLine Mechanical, Proline Mechanical, eLine Electronic