Serological pipettes
5 ml, 0,1 ml, +, blå, Individuelt pakket, 500/æske
Katalog # SARS86.1253.001
Marketsource
Leverandør: SARSTEDT
Gå til:
Product Details & Documents
PS, graduated, sterile.
- Plugged
- Easy identification by colour code
- Larger pipetting volume due to negative graduation
- Optimised mouth piece for universal fit in most common pipettors
- Easy to open, static-free packaging
Documents
Specifikationer
- Farvekode:blå
- Pakket:Individuelt pakket, 500/æske
- Steril:+
- Kapacitet:5 ml
- Inddeling:0,1 ml