96-Well micro test plates
Micro test plate with flat bottom, without lid, klar, 96, 25/bag
Katalog # SARS82.1581
Marketsource
Leverandør: SARSTEDT
Gå til:
Product Details & Documents
Clear PS
- Compatible with all common microplate dispensers, readers, and washers
- Excellent and consistent optical quality
- Optimal surface
- Uniform design
- Choice of flat, round or conical wells
Specifikationer
- Ant. brønde:96
- Pakket:25/bag
- Farve:klar
- Beskrivelse:Micro test plate with flat bottom, without lid
Accessories
Produktinformation
- Katalog #FarveTilgængelighedPrice
- Låg til mikrotestplader, PS