Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Containers, disposable, with screw cap

Containers, disposable, with screw cap

14 mm, for 100 ml containers, neutralt, 50/bag, 500/case, HDPE
Katalog # SARS76.564
Marketsource
Leverandør:   SARSTEDT
Containers, disposable, with screw cap
Katalog # SARS76.564
Leverandør:  SARSTEDT
Marketsource
Containers, disposable, with screw cap

14 mm, for 100 ml containers, neutralt, 50/bag, 500/case, HDPE

Price
242,32 kr
Log ind for tilgængelighed
1 * 500 Items
Product Details & Documents

PP, translucent, with neutral HDPE screw cap

  • Sterile or non-sterile
  • Without lid or with attached or enclosed lid available
  • Safety label ensures sterility prior to use and provides a convenient writing space

Ideal as general specimen containers.

Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904

Specifikationer
  • Pakket:
    50/bag, 500/case
  • Model:
    for 100 ml containers
  • Højde:
    14 mm
  • Materiale:
    HDPE
  • Lågfarve:
    neutralt