Pipette tips
Type J, 20 µl, -, Transparent, Eppendorf Reference, Research/Research Plus, mLine Mechanical, Proline Mechanical, eLine Electronic, 96/box, 1920/case
SARS70.1116.100
Marketsource
: SARSTEDT
These high quality tips are suitable for the most common pipettes.
- Sterile tips are DNase/RNase- and ATP-free, and non pyrogenic
- Available loose in bags, packed in racks or in Tip StackPack reload systems
Tip StackPack: Tips are preloaded in reusable, stacked 96-tip trays. The trays are designed to safely cover each individual 96-tip layer, shielding the preloaded tips in the stack like a protective screen.
- Pakket:96/box, 1920/case
- Steril:-
- Volumen:20 µl
- Seq:20
- Spidstype:Type J
- Spits farge:Transparent
- Til brug med:Eppendorf Reference, Research/Research Plus, mLine Mechanical, Proline Mechanical, eLine Electronic