Orders LinkContactUsLinkComponent
Vacuum Controller, V-850/V-855
Vacuum Controller, V-850/V-855
Vakuumstyreenhed V-850 til R-200/R-205
Katalog # 531-0233
Vacuum Controller, V-850/V-855
Katalog # 531-0233
Leverandør:  
Vacuum Controller, V-850/V-855
Vacuum Controller, V-850/V-855
Vakuumstyreenhed V-850 til R-200/R-205
Discontinued
Produktet er udgået. Men vi fandt lignende alternative produkter.
Alternativer
[EN]INTERFACE I-300 VACUBOX VALVEUNIT 1 * 1 Items Katalog #BUCH11I300S30
Produktet BUCH11I300S30 ligner 531-0233, men producenten og specifikationerne kan variere.
I-300 PRO VACUBOX VALVEUNIT 1 * 1 Items Katalog #BUCH11I300P30
Produktet BUCH11I300P30 ligner 531-0233, men producenten og specifikationerne kan variere.
Product Details & Documents
V-850 vacuum controller:
The V-850 vacuum controller with ¼" LCD VGA screen can be used universally for the display, control and adjustment of vacuum. The vacuum controller is a compact unit suitable for many tasks in the laboratory thanks to its integrated vent valve and chemical-resistant pressure sensor. It includes a timer, laboratory vacuum controller (Labvac) and solvent database featuring the 43 most commonly used solvents. The rotation speed control with the V-700 or V-710 vacuum pump enables accurate, hysteresis-free vacuum control with quiet operation.
Users also have the option of reducing their water consumption by adding a cooling water valve.
V-855 vacuum controller:
The V-855 vacuum controller comes with the same features as the V-850, and in addition: gradient and repetition functions for distillation in difficult conditions and the EasyVac function — automatic distillation based on vapour pressure detection.
The V-855 vacuum controller comes with the same features as the V-850, and in addition: gradient and repetition functions for distillation in difficult conditions and the EasyVac function — automatic distillation based on vapour pressure detection.
Leveringsinformation: Delivered with an air vent valve and pressure sensors.
Specifikationer
  • Beskrivelse:
    Vakuumstyreenhed V-850 til R-200/R-205
  • Seq:
    04
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Kommunikationskabel til R-210/R-215
    Kommunikationskabel til R-210/R-215
    181-0138
    Log ind for tilgængelighed
    1 * 1 Items - 279,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Woulff-flaske til V-850/V-855
    Woulff-flaske til V-850/V-855
    181-0141
    Log ind for tilgængelighed
    1 * 1 Items - 1.980,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Automatisk destillationsprobe til V-855
    Automatisk destillationsprobe til V-855
    531-0181
    Log ind for tilgængelighed
    1 * 1 Items - 2.090,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Monteringssæt til R-210/R-215
    Monteringssæt til R-210/R-215
    531-0236
    Log ind for tilgængelighed
    1 * 1 Items - 641,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Fjernstyring til V-850/V-855
    Fjernstyring til V-850/V-855
    531-0237
    Log ind for tilgængelighed
    1 * 1 Items - 5.590,00 kr