Disposal cabinet
Connecting frame for disposal cabinet/dispenser unit, 80×30×160 mm
Katalog # 134-0487
Leverandør: Palbam
Gå til:
Product Details & Documents
This disposal cabinet can be used as a stand-alone unit or in combination with the dispenser unit (connecting frame required).
- Made of stainless steel 304 with electropolished finish
- Two waste openings, two doors and levelling feet
- Does not require connecting to the wall
Can be used in cleanroom and critical environments.
Certifications: ISO certified.
Leveringsinformation: Typically available from stock for direct shipment from the supplier. Supplied with two large plastic bins.
Documents
Palbam Class - Catalog 2018
Produktlitteratur
Specifikationer
- Beskrivelse:Connecting frame for disposal cabinet/dispenser unit
- Seq:02
- B×D×H:80×30×160 mm