Orders LinkContactUsLinkComponent
Disposal cabinet
Disposal cabinet, 800×430×900 mm
Katalog # 134-0486
Leverandør:   Palbam
Disposal cabinet
Katalog # 134-0486
Leverandør:  Palbam
Disposal cabinet
Disposal cabinet
Disposal cabinet, 800×430×900 mm
Price
22.540,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
This disposal cabinet can be used as a stand-alone unit or in combination with the dispenser unit (connecting frame required).
  • Made of stainless steel 304 with electropolished finish
  • Two waste openings, two doors and levelling feet
  • Does not require connecting to the wall
Can be used in cleanroom and critical environments.
 : ISO certified.
 : Typically available from stock for direct shipment from the supplier. Supplied with two large plastic bins.
Specifikationer
  • Beskrivelse:
    Disposal cabinet
  • Seq:
    03
  • B×D×H:
    800×430×900 mm