Disposal cabinet
Disposal cabinet, 800×430×900 mm
Katalog # 134-0486
Leverandør: Palbam
Gå til:
Product Details & Documents
This disposal cabinet can be used as a stand-alone unit or in combination with the dispenser unit (connecting frame required).
- Made of stainless steel 304 with electropolished finish
- Two waste openings, two doors and levelling feet
- Does not require connecting to the wall
Can be used in cleanroom and critical environments.
: ISO certified.
: Typically available from stock for direct shipment from the supplier. Supplied with two large plastic bins.
Specifikationer
- Beskrivelse:Disposal cabinet
- Seq:03
- B×D×H:800×430×900 mm