Dråbefangere, Stutzer, lige
29/32 NS, 14/23 NS
201-1066
DURAN®, borosilicate 3.3 glass, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Anbringes direkte på destillationskolben for at fange dråber, med to standardslib.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
- Konusstr.:14/23 NS
- Kappestr.:29/32 NS
- Seq:02