Pipettespidser, universal, Axygen®
Eppendorf, graduated, 100 - 1000 µl, -, Blue, 12 racks, 96 each
Katalog # 613-1336
Leverandør: Corning
Gå til:
Product Details & Documents
Disse universelle pipettespidser er Kompatibel med de fleste mikropipetter.
- PP, autoclavable
- Smooth finish for minimum retention of collected sample
- Certified free of RNase, DNase and endotoxins
- Maxymum Recovery® tips feature ultra smooth surfaces: For reduced sample loss and maximal accuracy and precision
: Fås løse eller i racks, sterile eller usterile versioner.
Specifikationer
- Pakket:12 racks, 96 each
- Steril:-
- Volumen:100 - 1000 µl
- Seq:63
- Spidstype:Eppendorf, graduated
- Spits farge:Blue