Orders LinkContactUsLinkComponent
Glass beads
Glass beads
Glass beads, solid, 3 mm
Katalog # LENZ05124003
Leverandør:   Lenz Laborglas
Glass beads
Katalog # LENZ05124003
Leverandør:  Lenz Laborglas
Glass beads
Glass beads
Glass beads, solid, 3 mm
Price
285,00 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
Solid soda-lime glass.
Specifikationer
  • Ø:
    3 mm
  • Beskrivelse:
    Glass beads, solid
  • Seq:
    02