Funnel for conical ground joints
14/23 NS, 40 mm
Katalog # 221-1418
Leverandør: VITLAB
Gå til:
Product Details & Documents
PP, translucent.
- Ideal for transferring liquid or powdered reagents to multiple-neck flasks
- Very good chemical resistance
- Autoclavable
The funnel body is flattened on the side and the end of the shaft is shaped to fit conical ground joints.
Specifikationer
- Tragt O-Ø flad side:40 mm
- Stilk til:14/23 NS