Orders LinkContactUsLinkComponent
Funnel for conical ground joints
14/23 NS, 40 mm
Katalog # 221-1418
Leverandør:   VITLAB
Funnel for conical ground joints
Katalog # 221-1418
Leverandør:  VITLAB
Funnel for conical ground joints
Funnel for conical ground joints
14/23 NS, 40 mm
Price
437,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
PP, translucent.
  • Ideal for transferring liquid or powdered reagents to multiple-neck flasks
  • Very good chemical resistance
  • Autoclavable
The funnel body is flattened on the side and the end of the shaft is shaped to fit conical ground joints.
Specifikationer
  • Tragt O-Ø flad side:
    40 mm
  • Stilk til:
    14/23 NS