Orders LinkContactUsLinkComponent
Screw caps
Screw caps
GL 40
Katalog # VITL83315
Leverandør:   VITLAB
Screw caps
Katalog # VITL83315
Leverandør:  VITLAB
Screw caps
Screw caps
GL 40
Price
322,00 kr
Log ind for tilgængelighed
1 * 12 Items
Product Details & Documents
Made of PP, translucent.
Caps are autoclavable at 121 °C (2 bar) according to DIN EN 285.
Specifikationer
  • Gevind:
    GL 40