Screw caps
GL 25
Catalog # VITL83311
Supplier: VITLAB
Jump to:
Product Details & Documents
Made of PP, translucent.
Caps are autoclavable at 121 °C (2 bar) according to DIN EN 285.
Specifications
- Gevind:GL 25
Made of PP, translucent.
Caps are autoclavable at 121 °C (2 bar) according to DIN EN 285.