Screw caps
GL 18
Katalog # VITL83310
Leverandør: VITLAB
Gå til:
Product Details & Documents
Made of PP, translucent.
Caps are autoclavable at 121 °C (2 bar) according to DIN EN 285.
Specifikationer
- Gevind:GL 18