Funnel for conical ground joints
19/26 NS, 50 mm
Catalog # 221-1409
Supplier: VITLAB
Jump to:
Product Details & Documents
PP, translucent.
- Ideal for transferring liquid or powdered reagents to multiple-neck flasks
- Very good chemical resistance
- Autoclavable
The funnel body is flattened on the side and the end of the shaft is shaped to fit conical ground joints.
Specifications
- Tragt O-Ø flad side:50 mm
- Stilk til:19/26 NS