Orders LinkContactUsLinkComponent
Funnel for conical ground joints
Funnel for conical ground joints
19/26 NS, 50 mm
Catalog # 221-1409
Supplier:   VITLAB
Funnel for conical ground joints
Catalog # 221-1409
Supplier:  VITLAB
Funnel for conical ground joints
Funnel for conical ground joints
19/26 NS, 50 mm
Price
589,00 kr
Log in for availability
1 * 10 Items
Product Details & Documents
PP, translucent.
  • Ideal for transferring liquid or powdered reagents to multiple-neck flasks
  • Very good chemical resistance
  • Autoclavable
The funnel body is flattened on the side and the end of the shaft is shaped to fit conical ground joints.
Specifications
  • Tragt O-Ø flad side:
    50 mm
  • Stilk til:
    19/26 NS