Orders LinkContactUsLinkComponent
Tweezers
Tweezers
145 mm
Catalog # VITL68199
Supplier:   VITLAB
Tweezers
Catalog # VITL68199
Supplier:  VITLAB
Tweezers
Tweezers
145 mm
Price
22,50 kr
Log in for availability
1 * 1 Items
Product Details & Documents
POM, yellow. Universal tweezers for a variety of applications.
  • Blunt, elastic, very good resilience
  • With grooves on the outside for optimum handling and grip
  • Good chemical resistance
Specifications
  • Længde:
    145 mm