Watch dishes
Watch glass, PTFE, without base, 100 mm
Katalog # VITL113397
Leverandør: VITLAB
Gå til:
Product Details & Documents
Watch dishes, made of PTFE, white.
- Without base
- Very good chemical resistance
- Good temperature resistance
Ideal for use to cover beakers.
Certifications: DIN EN 285
Specifikationer
- Ø:100 mm
- Beskrivelse:Watch glass, PTFE, without base
- Seq:03