Orders LinkContactUsLinkComponent
Watch dishes
Watch dishes
Watch glass, PTFE, without base, 50 mm
Katalog # VITL113197
Leverandør:   VITLAB
Watch dishes
Katalog # VITL113197
Leverandør:  VITLAB
Watch dishes
Watch dishes
Watch glass, PTFE, without base, 50 mm
Price
142,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Watch dishes, made of PTFE, white.
  • Without base
  • Very good chemical resistance
  • Good temperature resistance
Ideal for use to cover beakers.
Certifications: DIN EN 285
Specifikationer
  • Ø:
    50 mm
  • Beskrivelse:
    Watch glass, PTFE, without base