Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Microtube racks

48 (4×12), 2,2 ml, 265×100×50 mm
Katalog # 211-8064
Microtube racks
Katalog # 211-8064
Microtube racks
Microtube racks

48 (4×12), 2,2 ml, 265×100×50 mm

Price
930,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Acrylic.

Specifikationer
  • Til rør:
    2,2 ml
  • Ant. huller:
    48 (4×12)
  • Seq:
    17
  • B×D×H:
    265×100×50 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.