Orders LinkContactUsLinkComponent
Racks for test tubes, acrylic
21 mm, 20 (2×10), — mm, — mm
  212-8361
Racks for test tubes, acrylic
  212-8361
Racks for test tubes, acrylic
Racks for test tubes, acrylic
21 mm, 20 (2×10), — mm, — mm
787,00 kr
1 * 1 Items
Made of acrylic, clear.
These test tube racks are of stable version, solid, hygienic, easily cleaned and physiologically inert with intermediate level, recesses in base.
  • Hul Ø:
    21 mm
  • Ant. huller:
    20 (2×10)
  • Højde:
    — mm
  • L×B:
    — mm
  • Seq:
    15