Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Microtube racks

Microtube racks

blå, 24 (2×12), 1,5 ml, 270×50×50 mm
Katalog # 212-6635
Microtube racks
Katalog # 212-6635
Microtube racks
Microtube racks

blå, 24 (2×12), 1,5 ml, 270×50×50 mm

Price
607,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Acrylic.

Specifikationer
  • Til rør:
    1,5 ml
  • Ant. huller:
    24 (2×12)
  • Farve:
    blå
  • Seq:
    05
  • B×D×H:
    270×50×50 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.