Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Microtube racks

Microtube racks

rød, 24 (2×12), 1,5 ml, 270×50×50 mm
Katalog # 212-6636
Microtube racks
Katalog # 212-6636
Microtube racks
Microtube racks

rød, 24 (2×12), 1,5 ml, 270×50×50 mm

Price
678,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Acrylic.

Specifikationer
  • Til rør:
    1,5 ml
  • Ant. huller:
    24 (2×12)
  • Farve:
    rød
  • Seq:
    06
  • B×D×H:
    270×50×50 mm
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.