Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Microtube racks

Microtube racks

orange, 48 (4×12), 1,5 ml, 270×100×50 mm
Catalog # 212-6637
Microtube racks
Catalog # 212-6637
Microtube racks
Microtube racks

orange, 48 (4×12), 1,5 ml, 270×100×50 mm

Price
1.110,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Acrylic.

Specifications
  • Til rør:
    1,5 ml
  • Ant. huller:
    48 (4×12)
  • Farve:
    orange
  • Seq:
    07
  • B×D×H:
    270×100×50 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.