Orders LinkContactUsLinkComponent
Vacuum aspiration systems
Vacuum aspiration system, with base plate, holder and 2 l bottle
Catalog # KOEHE-1550
Vacuum aspiration systems
Catalog # KOEHE-1550
Vacuum aspiration systems
Vacuum aspiration systems
Vacuum aspiration system, with base plate, holder and 2 l bottle
Price
14.340,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Vacuum aspiration system. A plastic supporting plate with carrying handles and attachments for holding the vacuum-proof suction flask make the system compact and mobile at any time.
Specifications
  • Beskrivelse:
    Vacuum aspiration system, with base plate, holder and 2 l bottle
Accessories
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 442-9468
    Foot switch with a 230 V socket
    Log in for availability
    1 * 1 Items - 2.860,00 kr