Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

UV mini lamps

UV-minilampe, kortbølge, med nylonstrop, 159 × 25 × 56 mm, UVG-4, Feltbrug, 254 nm, 4 W
Katalog # UVPA95-0158-04
UV mini lamps
Katalog # UVPA95-0158-04
UV mini lamps
UV mini lamps

UV-minilampe, kortbølge, med nylonstrop, 159 × 25 × 56 mm, UVG-4, Feltbrug, 254 nm, 4 W

Price
1.130,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

These lamps are excellent for quick field inspection or fluorescence applications.

  • For field use and fluorescence work
  • Longwave, shortwave, and combination options available
  • Four-watt intensity
  • Impact-resistant plastic

Mini UV Lamps feature four-watt intensity and are powered by four AA batteries (not included). Each lamp housing is made of impact-resistant plastic that resists fading or cracking. The lamps are lightweight and small enough to fit into a pocket. A nylon strap is attached for easy handling. The lamps are available in three configurations: longwave, shortwave, or a multi-band longwave/shortwave combination.

 : Model UVL-4F includes a flashlight and a longwave UV.

Specifikationer
  • Til:
    Feltbrug
  • Beskrivelse:
    UV-minilampe, kortbølge, med nylonstrop, 159 × 25 × 56 mm
  • Model:
    UVG-4
  • Udgang:
    4 W
  • Bølgelængde:
    254 nm
Accessories

Produktinformation

  • Katalog #
    Til
    Bølgelængde
    Tilgængelighed
    Price
  • Replacement shortwave UV tube
    Replacement shortwave UV tube
    733-2062
    UVG-11, UVGL-25
    254 nm
    Log ind for tilgængelighed
    1 * 1 Items - 340,00 kr

Produktinformation

  • Katalog #
    Til
    Bølgelængde
    Tilgængelighed
    Price
  • Replacement UV tube, longwave, BLB
    Replacement UV tube, longwave, BLB
    UVPA34-0010-01
    SL-2M
    365 nm
    Log ind for tilgængelighed
    1 * 1 Items - 258,00 kr