Pipette tips
Bevelled, non-graduated, 200 - 1000 µl, -, 76 mm, blå, 96 tips/rack; 6 racks/case
Katalog # ALPCLW6470
Leverandør: Alpha Laboratories
Gå til:
Product Details & Documents
These tips are manufactured from medical grade PP in modern production facilities with stringent quality controls at every stage.
- Tips are lot tested and certified for absence of detectable human DNA and detectable endotoxins (pyrogens)
- Tips are tested and certified for absence of heavy metals, detectable DNase and RNase
- With graduations to visually check volume
- Suitable for all major pipettor brands
Specifikationer
- Farvekode:blå
- Pakket:96 tips/rack; 6 racks/case
- Steril:-
- Volumen:200 - 1000 µl
- Længde:76 mm
- Seq:09
- Spidstype:Bevelled, non-graduated
- Cat. No.:ALPCLW6470