Orders LinkContactUsLinkComponent
RNase A
RNase A
5 mL
Katalog # AC118
Leverandør:   OMEGA BIO-TEK
RNase A
Katalog # AC118
Leverandør:  OMEGA BIO-TEK
CAS nummer:  
CAS nummer:  
Price
1.510,00 kr
Log ind for tilgængelighed
1 * 5 mL
Product Details & Documents
Ribonuclease A (RNase A) is endonuclease-free and quality controlled for use in plasmid purification procedures for digestion of RNA.
Specifikationer
Specifikationer
  • Pakn:
    5 mL
  • Cat. No.:
    AC118
  • Koncentration:
    25 mg/ml
  • Enzym navn:
    RNase A