Clamps for universal platforms
250 ml kolbeklemme, 9 (280×330); 16 (457×457); 24 (457×609) for MaxQ 2000
24 (760×457) and 40 (910×609) clamps for MaxQ 3000
24 (760×457) and 40 (910×609) clamps for MaxQ 3000
Katalog # 444-0308
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Documents
BARN30156BI_TDS_EN
Datablad
Specifikationer
- Til:9 (280×330); 16 (457×457); 24 (457×609) for MaxQ 2000
24 (760×457) and 40 (910×609) clamps for MaxQ 3000 - Beskrivelse:250 ml kolbeklemme



