Clamps for universal platforms
50 ml kolbeklemme, 30 (280×330); 32 (457×457); 40 (457×609) for MaxQ 2000
112 (760×457) and 160 (910×609) clamps for MaxQ 3000
112 (760×457) and 160 (910×609) clamps for MaxQ 3000
Katalog # 444-0306
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Documents
BARN30156BI_TDS_EN
Datablad
Specifikationer
- Til:30 (280×330); 32 (457×457); 40 (457×609) for MaxQ 2000
112 (760×457) and 160 (910×609) clamps for MaxQ 3000 - Beskrivelse:50 ml kolbeklemme
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



